missing translation for 'onlineSavingsMsg'
Learn More

Bis(carboxyethylgermanium) sesquioxide, 99.7%

Product Code. 11324277
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324277 25 g 25g
11314277 5 g 5g
11304277 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324277 Supplier Thermo Scientific Alfa Aesar Supplier No. 036758.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12758-40-6 | C6H10Ge2O7 | 339.399 g/mol

A bifunctional metalloligand with metal-C (Ge-C) bonds,bis(carboxyethylgermanium) sesquioxide, has been used to make two metal-organometallic frameworks (MOMFs). bis-(carboxyethylgermanium)sesquioxide is a as a promising anti-oxidative agent. Bis(carboxyethylgermanium) sesquioxide induce INF-γ and has undergone clinical trials with promising preclinical results.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12758-40-6
Molecular Formula C6H10Ge2O7
Molecular Weight (g/mol) 339.399
MDL Number MFCD00049319
InChI Key XEABSBMNTNXEJM-UHFFFAOYSA-N
Synonym propagermanium, proxigermanium, carboxyethylgermanium sesquioxide, 14c-proxigermanium, 2-carboxyethylgermasesquioxane, dipropanoic acid germanium sequioxide, 2-carboxyethyl germanium sesquioxide, bis 2-carboxyethylgermanium sesquioxide, ccris 5469, carboxylethylgermanium sesquioxide
PubChem CID 83030
ChEBI CHEBI:32060
IUPAC Name 3-[[2-carboxyethyl(oxo)germyl]oxy-oxogermyl]propanoic acid
SMILES C(C[Ge](=O)O[Ge](=O)CCC(=O)O)C(=O)O

Specifications

Melting Point 320°C (decomposition)
Odor Odorless
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 339.25
Percent Purity 99.7%
Physical Form Powder
Chemical Name or Material Bis(carboxyethylgermanium) sesquioxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.