missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)ruthenium, Ru 43.2% min

Product Code. 11367547
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11367547

Brand: Thermo Scientific Alfa Aesar 012781.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1287-13-4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1287-13-4
MDL Number MFCD00001442
Synonym Ruthenocene

Specifications

Melting Point 194°C to 198°C
Quantity 5 g
Sensitivity Air and moisture sensitive
Formula Weight 231.26
Concentration or Composition (by Analyte or Components) Ru 43.2% min.
Physical Form Crystalline
Chemical Name or Material Bis(cyclopentadienyl)ruthenium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.