Learn More
Bis(cyclopentadienyl)titanium dichloride, Thermo Scientific™
20.90 €
Chemical Identifiers
| CAS | 1271-19-8 |
|---|---|
| Molecular Formula | C10H10Cl2Ti-2 |
| Molecular Weight (g/mol) | 248.957 |
| MDL Number | MFCD00003723 |
| InChI Key | MKNXBRLZBFVUPV-UHFFFAOYSA-L |
| Synonym | Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride |
| PubChem CID | 124040768 |
| IUPAC Name | cyclopenta-1,3-diene;titanium(2+);dichloride |
| SMILES | [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under argon
Chemical Identifiers
| 1271-19-8 | |
| 248.957 | |
| MKNXBRLZBFVUPV-UHFFFAOYSA-L | |
| 124040768 | |
| [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] |
| C10H10Cl2Ti-2 | |
| MFCD00003723 | |
| Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride | |
| cyclopenta-1,3-diene;titanium(2+);dichloride |
Specifications
| 1271-19-8 | |
| Odorless | |
| MFCD00003723 | |
| 14,9482 | |
| It is insoluble in water | |
| [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2] | |
| 248.957 | |
| 249 |
| 1.60 g/mL | |
| C10H10Cl2Ti-2 | |
| Air and moisture sensitive | |
| Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride | |
| MKNXBRLZBFVUPV-UHFFFAOYSA-L | |
| cyclopenta-1,3-diene;titanium(2+);dichloride | |
| 124040768 | |
| Bis(cyclopentadienyl)titanium dichloride |
Safety and Handling
GHS H Statement
H341-H302-H332-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 215-035-9
missing translation for 'rtecsNumber' : XR2050000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures; Store under Argon
- Bis(cyclopentadienyl)titanium dichloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.