missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 012547.06


Quantity 5g

 View more versions of this product

Product Code. 11357387

Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1271-19-8
248.957
MKNXBRLZBFVUPV-UHFFFAOYSA-L
124040768
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
C10H10Cl2Ti-2
MFCD00003723
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
cyclopenta-1,3-diene;titanium(2+);dichloride

Specifications

1271-19-8
Odorless
MFCD00003723
Air and moisture sensitive
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
MKNXBRLZBFVUPV-UHFFFAOYSA-L
cyclopenta-1,3-diene;titanium(2+);dichloride
124040768
Bis(cyclopentadienyl)titanium dichloride
1.60 g/mL
C10H10Cl2Ti-2
5g
14,9482
It is insoluble in water
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
248.957
249
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bis(cyclopentadienyl)titanium dichloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.