missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)zirconium dichloride, 98+%

Product Code. 10589253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10589253 25 g Glass bottle 25g
10052430 5 g Glass bottle 5g
10790965 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10589253 Supplier Thermo Scientific Acros Supplier No. 187920250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1291-32-3 | C10H10Cl2Zr | 292.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1291-32-3
Molecular Formula C10H10Cl2Zr
Molecular Weight (g/mol) 292.32
MDL Number MFCD00003726
Synonym Zirconocene dichloride

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 242.0°C to 245.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Fieser 11,51; 16,393
Solubility Information Solubility in water: hydrolysis.
Formula Weight 292.32
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Bis(cyclopentadienyl)zirconium dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.