missing translation for 'onlineSavingsMsg'
Learn More

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, 95%

Product Code. 12685117 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12685117 5 g Glass bottle 5g
12675117 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12685117 Supplier Thermo Scientific Acros Supplier No. 447890050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 887919-35-9 | C32H56Cl2N2P2Pd | 708.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887919-35-9
Molecular Formula C32H56Cl2N2P2Pd
Molecular Weight (g/mol) 708.08
MDL Number MFCD09265123
InChI Key DWOZNANUEDYIOF-UHFFFAOYSA-L
Synonym bis di-tert-butyl 4-dimethylaminophenyl phosphine dichloropalladium ii, pd amphos cl2, pdcl2 amphos 2, a-taphos 2pdcl2, dichlorobis di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii, alpha-taphos 2pdcl2, bis 4-di-tert-butylphosphino-n,n-dimethylbenzenamine palladium dichloride
PubChem CID 11714597
IUPAC Name 4-ditert-butylphosphanyl-N,N-dimethylaniline;dichloropalladium
SMILES Cl[Pd]Cl.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C

Specifications

Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 708.07
Percent Purity 95%
Chemical Name or Material Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium
compliance-icons
Product Identifier
  • Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.