missing translation for 'onlineSavingsMsg'
Learn More

Bis(dibenzylideneacetone)palladium

Product Code. 10571511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571511 1 g Glass bottle 1g
10678201 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10571511 Supplier Thermo Scientific Acros Supplier No. 291970010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32005-36-0 | C34H28O2Pd | 575.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Molecular Weight (g/mol) 575.02
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 150.0°C
Color Black to Brown-Red
Packaging Glass bottle
Linear Formula Pd(C6H5CH=CHC(O)CH=CHC6H5)2
Quantity 1 g
Fieser 15,302
Solubility Information Solubility in water: insoluble.
Formula Weight 575.02
Percent Purity 16 to 21% (Pd)
Physical Form Fine Powder
Chemical Name or Material Bis(dibenzylideneacetone)palladium
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(dibenzylideneacetone)palladium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.