missing translation for 'onlineSavingsMsg'
Learn More

Bis(dichlorophosphino)methane, 96%

Product Code. 10494505 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10494505

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 28240-68-8 | CH2Cl4P2 | 217.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28240-68-8
Molecular Formula CH2Cl4P2
Molecular Weight (g/mol) 217.79
MDL Number MFCD01073767
InChI Key OEKLCAXLANZKPX-UHFFFAOYSA-N
Synonym bis dichlorophosphino methane, dichloro dichlorophosphanyl methyl phosphane, acmc-1cq46, methylenebis dichlorophosphine, methylenebis phosphonous dichloride, dichloro dichlorophosphanylmethyl phosphane, phosphonous dichloride,p,p'-methylenebis
PubChem CID 2784185
IUPAC Name dichloro(dichlorophosphanylmethyl)phosphane
SMILES C(P(Cl)Cl)P(Cl)Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6010g/mL
Boiling Point 101.0°C to 105.0°C (22.0 mmHg)
Flash Point 96°C
Assay Percent Range 95% min. (Argentometry)
Packaging Glass bottle
Linear Formula (Cl2P)2CH2
Refractive Index 1.5920 to 1.5950
Quantity 1 g
Specific Gravity 1.601
Solubility Information Solubility in water: reacts
Formula Weight 217.79
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bis(dichlorophosphino)methane
Show More Show Less
compliance-icons
Product Identifier
  • Bis(dichlorophosphino)methane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.