missing translation for 'onlineSavingsMsg'
Learn More

Bis(diisopropylamino)chlorophosphine

Product Code. 10135620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10135620 5 g Glass bottle 5g
10627684 1 g Glass bottle 1g
11995841 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10135620 Supplier Thermo Scientific Acros Supplier No. 321560050
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 56183-63-2 | C12H28ClN2P | 266.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56183-63-2
Molecular Formula C12H28ClN2P
Molecular Weight (g/mol) 266.79
MDL Number MFCD00061482
InChI Key FEHUTHGOLLQBNW-UHFFFAOYSA-N
Synonym bis diisopropylamino chlorophosphine, phosphorodiamidous chloride, tetrakis 1-methylethyl, chloro diisopropylamino phosphanyl diisopropylamine, pubchem6478, acmc-1av77, bisdiisopropylaminochlorophosphine, bis diisopropyamino chlorophosphine, bis di-isopropylamino chlorophosphine, bis n,n-diisopropylamino chlorophosphine, chlorobis n,n-diisopropylamino phosphine
PubChem CID 2733425
IUPAC Name N-[chloro-[di(propan-2-yl)amino]phosphanyl]-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(Cl)N(C(C)C)C(C)C

Specifications

Melting Point 100.0°C to 110.0°C
Packaging Glass bottle
Linear Formula [((CH3)2CH)2N]2PCl
Quantity 5 g
Solubility Information Solubility in water: reacts
Formula Weight 266.8
Chemical Name or Material Bis(diisopropylamino)chlorophosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.