missing translation for 'onlineSavingsMsg'
Learn More

Bis(dimethylthiocarbamyl) Sulfide 97%, ACROS Organics™

Product Code. 10537362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
250g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10537362 250g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10537362 Supplier Thermo Scientific Acros Supplier No. 402432500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 97-74-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 106.0°C to 110.0°C
Color Yellow
Flash Point 156°C
Loss on Drying 0.3% max. (60°C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H12N2S3
Linear Formula ((CH3)2NCS)2S
MDL Number MFCD00014870
Quantity 250 g
Synonym tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm
InChI Key REQPQFUJGGOFQL-UHFFFAOYSA-N
SMILES CN(C)C(=S)SC(=S)N(C)C
IUPAC Name dimethylcarbamothioyl N,N-dimethylcarbamodithioate
Molecular Weight (g/mol) 208.356
PubChem CID 7347
Formula Weight 208.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Bis(dimethylthiocarbamyl) sulfide, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(dimethylthiocarbamyl) sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.