missing translation for 'onlineSavingsMsg'
Learn More

Bis(methylcyclopentadienyl)niobium dichloride, 97%, Thermo Scientific Chemicals

Product Code. 11498580
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498580 1 g 1g
11488580 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11498580 Supplier Thermo Scientific Alfa Aesar Supplier No. 044393.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61374-51-4
Molecular Formula C12H14Cl2Nb
MDL Number MFCD03427138
Synonym bis methylcyclopentadienyl niobium dichloride, acmc-1b8dp, bis 2-methylcyclopenta-1,3-dien-1-yl niobiumbis ylium dichloride, niobium,dichlorobis 1,2,3,4,5-h-1-methyl-2,4-cyclopentadien-1-yl

Specifications

Melting Point 180°C (decomposition)
Color Brown
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Formula Weight 322.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Bis (methylcyclopentadienyl)niobium dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.