missing translation for 'onlineSavingsMsg'
Learn More

Bis(methylthio)methane, 99%

Product Code. 11458597
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458597 25 g 25g
11448597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458597 Supplier Thermo Scientific Alfa Aesar Supplier No. A19706.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1618-26-4 | C3H8S2 | 108.217 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1618-26-4
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.217
MDL Number MFCD00008564
InChI Key LOCDPORVFVOGCR-UHFFFAOYSA-N
Synonym bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
PubChem CID 15380
IUPAC Name bis(methylsulfanyl)methane
SMILES CSCSC

Specifications

Density 1.059
Boiling Point 146°C to 147°C
Flash Point 43°C (109°F)
Odor Unpleasant
Refractive Index 1.534
Quantity 25 g
UN Number UN1993
Beilstein 1731143
Merck Index 14,1256
Formula Weight 108.23
Percent Purity 99%
Chemical Name or Material Bis(methylthio)methane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.