missing translation for 'onlineSavingsMsg'
Learn More

Bis(neopentyl glycolato)diboron, 97%, Thermo Scientific Chemicals

Product Code. 11369455
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369455 250 mg 250mg
11349455 1 g 1g
11359455 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369455 Supplier Thermo Scientific Alfa Aesar Supplier No. L18675.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201733-56-4
Molecular Formula C10H20B2O4
Molecular Weight (g/mol) 225.89
MDL Number MFCD02093062
InChI Key MDNDJMCSXOXBFZ-UHFFFAOYSA-N
Synonym bis neopentyl glycolato diboron, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi 1,3,2-dioxaborinane, 2-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl-2,2'-bi-1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl, bis neopentyl glycolato diborane, bis 2,2-dimethyl-1,3-propanediolato diboron, 5,5,5',5'-tetramethyl-2,2' bi 1,3,2 dioxaborinanyl
PubChem CID 2734316
IUPAC Name 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES CC1(C)COB(OC1)B1OCC(C)(C)CO1

Specifications

Melting Point 170°C to 178°C
Quantity 250 mg
Beilstein 8839038
Formula Weight 225.89
Percent Purity 97%
Chemical Name or Material Bis(neopentyl glycolato)diboron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.