missing translation for 'onlineSavingsMsg'
Learn More

Bis(neopentyl glycolato)diboron, TRC

Product Code. 30331248
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331248 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331248 Supplier Toronto Research Chemicals Supplier No. TRCB50480010G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(neopentyl glycolato)diboron
CAS 201733-56-4
Molecular Formula C10H20B2O4
Quantity 10 g
Synonym 5,5,5',5'-Tetramethyl-2,2'-bi-1,3,2-dioxaborinane, Bis(neopentanediolato)diboron
InChI Formula InChI=1S/C10H20B2O4/c1-9(2)5-13-11(14-6-9)12-15-7-10(3,4)8-16-12/h5-8H2,1-4H3
SMILES CC1(C)COB(OC1)B2OCC(C)(C)CO2
IUPAC Name 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Weight (g/mol) 225.89
Formula Weight 226.1548
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.