missing translation for 'onlineSavingsMsg'
Learn More

Bis(norbornadiene)rhodium(I) tetrafluoroborate, 94%

Product Code. 10576173 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10576173 100 mg Glass bottle 100mg
10331763 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10576173 Supplier Thermo Scientific Acros Supplier No. 382551000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36620-11-8 | C14H16BF4Rh | 373.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36620-11-8
Molecular Formula C14H16BF4Rh
Molecular Weight (g/mol) 373.99
MDL Number MFCD00671775
InChI Key HAYDJWBQWOEERB-UHFFFAOYSA-N
Synonym bis norbornadiene rhodium i tetrafluoroborate, rh nbd 2bf4, bis eta-2,5-norbornadiene rhodium i tetrafluoroborate, bis norbornadiene tetrafluoroborato rhodium, bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate, rh nbd 2 bf4, bis norbomadiene rhodium i tetrafluoroborate, bis norbornadiene rhodium tetrafluoroborate
PubChem CID 10915722
IUPAC Name bicyclo[2.2.1]hepta-2,5-diene;rhodium;tetrafluoroborate
SMILES [Rh+].F[B-](F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2

Specifications

Melting Point 209.0°C to 211.0°C
Color Red
Elemental Analysis 92% min.
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: decomposes
Formula Weight 373.99
Percent Purity 94%
Physical Form Powder
Chemical Name or Material Bis(norbornadiene)rhodium(I) tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Bis(norbornadiene)rhodium(I) tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.