missing translation for 'onlineSavingsMsg'
Learn More

Bis(p-tolyl)phosphine oxide, 98%

Product Code. 15408068
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408068 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15408068 Supplier Thermo Scientific Alfa Aesar Supplier No. H61504.06
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2409-61-2 | C14H14OP+ | 229.239 g/mol

Bis(p-tolyl)phosphine oxide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(p-tolyl)phosphine oxide is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2409-61-2
Molecular Formula C14H14OP+
Molecular Weight (g/mol) 229.239
MDL Number MFCD01445489
InChI Key ZHIPXAFNKGZMSC-UHFFFAOYSA-N
Synonym bis p-tolyl phosphine oxide, di-p-tolylphosphine oxide, bis 4-methylphenyl phosphine oxide, di p-tolyl phosphine oxide, phosphine oxide, bis 4-methylphenyl, 1-methyl-4-4-methylphenylphosphoroso benzene, bis-p-tolylphosphine oxide, oxo bis-p-tolyl phosphonium, bis 4-tolyl phosphine oxide, ksc916e3n
PubChem CID 13357841
IUPAC Name bis(4-methylphenyl)-oxophosphanium
SMILES CC1=CC=C(C=C1)[P+](=O)C2=CC=C(C=C2)C

Specifications

Quantity 5 g
Beilstein 2804632
Solubility Information Slightly soluble in water.
Formula Weight 230.29
Percent Purity 98%
Chemical Name or Material Bis(p-tolyl)phosphine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.