missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)zirconium dichloride, 99%, Thermo Scientific Chemicals

Product Code. 11338368
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338368 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11338368 Supplier Thermo Scientific Alfa Aesar Supplier No. 039555.04
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54039-38-2
Molecular Formula C20H30Cl2Zr-2
Molecular Weight (g/mol) 432.584
MDL Number MFCD00058849
InChI Key OCFSLQKTBFSWPL-UHFFFAOYSA-L
Synonym decamethylzirconocene dichloride
PubChem CID 57369741
IUPAC Name dichlorozirconium;1,2,3,4,5-pentamethylcyclopenta-1,3-diene
SMILES C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.Cl[Zr]Cl

Specifications

Color Yellow
Packaging Ampule
Quantity 2 g
UN Number UN3096
Sensitivity Air and moisture sensitive
Formula Weight 432.56
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Bis(pentamethylcyclopentadienyl)zirconium dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.