missing translation for 'onlineSavingsMsg'
Learn More

Bis(pinacolato)diboron, TRC

Product Code. 30180383
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180383 25 g 25g
30180119 10 g 10g
30179766 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180383 Supplier Toronto Research Chemicals Supplier No. TRCB52277025G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73183-34-3
Molecular Formula C12H24B2O4
Molecular Weight (g/mol) 253.94
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bis(1,3,2-dioxaborolane), 4,4',5,5'-Octamethyl-2,2'-bi-1,3,2-dioxaborolane, B2Pin2, Bis(pinacolate)diboron, Bis(pinacolato) diborate, Bis(pinacolato)diborane, Diboron Pinacol Ester, Dipinacoldiboron, Pinacol Diborane;
IUPAC Name 4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)B2OC(C)(C)C(C)(C)O2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H24B2O4/c1-9(2)10(3,4)16-13(15-9)14-17-11(5,6)12(7,8)18-14/h1-8H3
Chemical Name or Material Bis(pinacolato)diboron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.