missing translation for 'onlineSavingsMsg'
Learn More

Bis(pyridine)iodonium tetrafluoroborate, 97%

Product Code. 10218753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10218753 5 g Glass bottle 5g
10723993 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10218753 Supplier Thermo Scientific Acros Supplier No. 369680050
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 15656-28-7 | C10H10BF4IN2 | 371.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15656-28-7
Molecular Formula C10H10BF4IN2
Molecular Weight (g/mol) 371.91
MDL Number MFCD03703393
InChI Key BMDSRCBKJZCUBH-UHFFFAOYSA-N
Synonym bis pyridine iodonium tetrafluoroborate, barluenga's reagent, bis pyridine iodonium i tetrafluoroborate, iodanium pyridine tetrafluoroborate, iodonium pyridine tetrafluoroborate, bispyridine iodonium tetrafluoroborate, bis pyridin iodonium tetrafluoroborate, iodium bis pyridine tetrafluoroborate, bis pyridine-iodonium tetrafluoroborate
PubChem CID 10883201
IUPAC Name iodanium;pyridine;tetrafluoroborate
SMILES F[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1

Specifications

Melting Point 137.0°C to 141.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 1303
Formula Weight 371.91
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Bis(pyridine)iodonium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Bis(pyridine)iodonium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.