missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-n-butyltin) Oxide, Stabilized 96%, Thermo Scientific™

Product Code. 10000610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10000610 5 mL Glass Bottle 5mL
10539810 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10000610 Supplier Thermo Scientific Acros Supplier No. 106510050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-35-9
Molecular Formula C24H58OSn2
Molecular Weight (g/mol) 600.15
MDL Number MFCD00009418
InChI Key NZQFYJHTWVEUPP-UHFFFAOYSA-N
Synonym tributyltin oxide, tbto, bis tributyltin oxide, hexabutyldistannoxane, distannoxane, hexabutyl, lastanox q, bis tri-n-butyltin oxide, butinox, biomet
PubChem CID 16682746
ChEBI CHEBI:81543
IUPAC Name tributyltin oxide
SMILES O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC

Specifications

Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 180.0°C (2.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Linear Formula [CH3(CH2)3]3SnOSn[(CH2)3CH3]3
Refractive Index 1.4854 to 1.4874
Quantity 5 mL
Fieser 06,56; 07,26; 08,43; 09,53; 11,62; 13,41; 16,227
Stabilizer 0.1% lactic acid
Formula Weight 596.11
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bis(tri-n-butyltin) oxide, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.