missing translation for 'onlineSavingsMsg'
Learn More

Bis(tributyltin) Oxide (Technical Grade), TRC

Product Code. 30337345
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337345 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337345 Supplier Toronto Research Chemicals Supplier No. TRCB59665550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBTO
CAS 56-35-9
Molecular Formula C24 H54 O Sn2
Quantity 50 g
Synonym TBTO (Bis(tributyltin) oxide), Distannoxane, 1,1,1,3,3,3-hexabutyl- (ACI), 1,1,1,3,3,3-Hexabutyldistannoxane (ACI), Bis(tributyltin) oxide (6CI), Distannoxane, hexabutyl- (8CI, 9CI), 6-Oxa-5,7-distannaundecane, 5,5,7,7-tetrabutyl-, Aceto TBTO, BioMeT 66, BioMeT SRM, BioMeT TBTO, Bis(tributylstannic oxide), Bis(tributylstannyl) ether, Bis(tributylstannyl) oxide, Butinox, Di-tributyl tin oxide, Diall, Hexabutyldistannoxane, Hexabutylditin oxide, Intercide 340A, Lastanox F, Lastanox Q, Lastanox Q1, Lastanox Super, Lastanox T, Lastanox T 20, MT 1E, Mykolastanox F, NSC 22332, NSC 28132, Oxybis(tributylstannane), Oxybis[tributyltin], Stannicide A, Stannicide O, TBTO, TnBTO, Tri-n-butyltin oxide, Tributyltin oxide, Vikol AF 25, Vikol LO 25
InChI Formula InChI=1S/6C4H9.O.2Sn/c6*1-3-4-2;;;/h6*1,3-4H2,2H3;;;
SMILES CCCC[Sn](CCCC)(CCCC)O[Sn](CCCC)(CCCC)CCCC
IUPAC Name tributyl(tributylstannyloxy)stannane
Molecular Weight (g/mol) 596.1
Formula Weight 598.2219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.