missing translation for 'onlineSavingsMsg'
Learn More

Bis(tricyclohexylphosphine)palladium(0), 97% min

Product Code. 15407075
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407075 250 mg 250mg
15402457 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407075 Supplier Thermo Scientific Alfa Aesar Supplier No. 045792.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Air Sensitive | CAS: 33309-88-5 | C36H66P2Pd | 667.29 g/mol

Bis(tricyclohexylphosphine)palladium(0) is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33309-88-5
Molecular Formula C36H66P2Pd
Molecular Weight (g/mol) 667.29
MDL Number MFCD01073796
InChI Key JGBZTJWQMWZVNX-UHFFFAOYSA-N
Synonym bis tricyclohexylphosphine palladium 0, bis tricyclohexylphosphine palladium o, bis tricyclohexylphosphine palladium, palladium, bis tricyclohexylphosphine, pd pcy3 2, palladium; tricyclohexylphosphane, palladium-tricyclohexylphosphine 1:2, bis tricyclohexylphosphine-palladium o
PubChem CID 2734559
IUPAC Name palladium;tricyclohexylphosphane
SMILES [Pd].C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Color White
Quantity 250 mg
Sensitivity Air and moisture sensitive
Solubility Information Soluble in chloroform,toluene. Insoluble in alcohol and water.
Formula Weight 667.29
Physical Form Crystalline
Chemical Name or Material Bis(tricyclohexylphosphine)palladium(0)
compliance-icons
Product Identifier
  • Bis(tricyclohexylphosphine)palladium(0)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bis(tricyclohexylphosphine)palladium(0)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.