missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)carbodiimide, 98%

Product Code. 10736573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10736573 25 g Glass bottle 25g
10006862 1 g Glass bottle 1g
10527323 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10736573 Supplier Thermo Scientific Acros Supplier No. 375130250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1000-70-0 | C7H18N2Si2 | 186.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000-70-0
Molecular Formula C7H18N2Si2
Molecular Weight (g/mol) 186.4
MDL Number MFCD00051538
InChI Key KSVMTHKYDGMXFJ-UHFFFAOYSA-N
Synonym bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
PubChem CID 70473
IUPAC Name N,N'-bis(trimethylsilyl)methanediimine
SMILES C[Si](C)(C)N=C=N[Si](C)(C)C

Specifications

Melting Point -23°C
Color Beige to Yellow or White
Density 0.8210g/mL
Boiling Point 164°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.435
Quantity 25 g
Specific Gravity 0.821
Solubility Information Solubility in water: insoluble
Formula Weight 186.4
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Bis(trimethylsilyl)carbodiimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.