missing translation for 'onlineSavingsMsg'
Learn More

Bisabolene, mixture of isomers

Product Code. 11482998
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11482998

Brand: Thermo Scientific Alfa Aesar A18724.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 495-62-5 | C15H24 | 204.357 g/mol

It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds use in artificial oil of bergamot, myrrh and lemon, and is an excellent fixative for neroli bases.

Solubility
Soluble in alcohol, water, (0.008994 mg/L @ 25°C (est)).

Notes
Air Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-62-5
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00129080
InChI Key XBGUIVFBMBVUEG-CCEZHUSRSA-N
Synonym e-gamma-bisabolene, unii-l8m7r52o3b, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, e, 1e-bisabola-1 10 ,4,7 11-triene 4e-4-1,5-dimethylhex-4-en-1-ylidene-1-methylcyclohexene, gamma-bisabolene, e, gamma-bisabolene, 4e, cyclohexene, 4-1,5-dimethyl-4-hexenylidene-1-methyl-, 4e, .gamma.-bisabolene, e gamma-bisabolene, 1-methyl-4-6-methylhept-5-en-2-ylidene cyclohex-1-ene
PubChem CID 3033866
ChEBI CHEBI:49238
IUPAC Name (4Z)-1-methyl-4-(6-methylhept-5-en-2-ylidene)cyclohexene
SMILES CC1=CCC(=C(C)CCC=C(C)C)CC1

Specifications

Density 0.89
Refractive Index 1.494
Quantity 50 g
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol,water,(0.008994mg/L @ 25°C (est)).
Formula Weight 204.36
Chemical Name or Material Bisabolene, mixture of isomers

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.