missing translation for 'onlineSavingsMsg'
Learn More

Bisindolylmaleimide I, MedChemExpress

Product Code. 16443396
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443396 25 mg 25mg
16435898 10 mg 10mg
16432047 50 mg 50mg
16428537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443396 Supplier Medchem Express Supplier No. HY13867
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bisindolylmaleimide I (GF109203X) is a highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor with a Ki of 14 nM.

Bisindolylmaleimide I (GF109203X) is a highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor with a Ki of 14 nM.

Specifications

Chemical Name or Material Bisindolylmaleimide I
CAS 133052-90-1
Color Red
Purity Grade Notes Research
Molecular Formula C25H24N4O2
Quantity 25 mg
Synonym GF109203X Go 6850
Solubility Information DMSO : ≥ 32 mg/mL (77.58 mM)
SMILES O=C(C(C1=CN(CCCN(C)C)C2=C1C=CC=C2)=C3C4=CNC5=C4C=CC=C5)NC3=O
Molecular Weight (g/mol) 412.48
Formula Weight 412.48
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.