missing translation for 'onlineSavingsMsg'
Learn More

Bismuth, 99%, powder, -100 mesh, Thermo Scientific Chemicals

Product Code. 10165520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10165520 100 g 100g
10721365 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10165520 Supplier Thermo Scientific Acros Supplier No. 318091000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Color Black
Melting Point 271.0°C
Boiling Point 1500.0°C
Density 9.8000g/mL
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 99.5%
Form Powder
Linear Formula BI
Fieser 13,39
Merck Index 15, 1261
Formula Weight 208.98
Percent Purity 99.5%
Chemical Name or Material Bismuth, 99.5%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.