Learn More
Bismuth(III) bromide, anhydrous, 99.999% (metals basis), Thermo Scientific™
100.00 € - 289.00 €
Chemical Identifiers
| CAS | 7787-58-8 |
|---|---|
| Molecular Formula | BiBr3 |
| Molecular Weight (g/mol) | 448.692 |
| MDL Number | MFCD00014195 |
| InChI Key | TXKAQZRUJUNDHI-UHFFFAOYSA-K |
| Synonym | bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo |
| PubChem CID | 82232 |
| IUPAC Name | tribromobismuthane |
| SMILES | Br[Bi](Br)Br |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15417445
|
Thermo Scientific Alfa Aesar
047175.03 |
1g |
100.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15427445
|
Thermo Scientific Alfa Aesar
047175.06 |
5g |
289.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| BiBr3 | |
| MFCD00014195 | |
| bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo | |
| tribromobismuthane |
Specifications
| 7787-58-8 | |
| MFCD00014195 | |
| bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo | |
| TXKAQZRUJUNDHI-UHFFFAOYSA-K | |
| tribromobismuthane | |
| 82232 | |
| 99.999% | |
| Hygroscopic | |
| (metals basis) |
| BiBr3 | |
| UN3260 | |
| Decomposes in water. | |
| Br[Bi](Br)Br | |
| 448.692 | |
| 448.71 | |
| Acidic | |
| 5.72 g/mL | |
| Bismuth(III) bromide, Anhydrous |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 232-121-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Bismuth(III) bromide, anhydrous
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.