missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride, 97+%, anhydrous

Product Code. 10512231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10512231 100 g Glass bottle 100g
10328490 25 g Glass bottle 25g
10308663 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10512231 Supplier Thermo Scientific Acros Supplier No. 208831000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-60-2
Molecular Formula BiCl3
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Spezifikation

Boiling Point 447.0°C
Melting Point 230.0°C to 232.0°C
Color Beige to White or Gray
Physical Form Crystalline Powder and/or Chunks
Quantity 100 g
Packaging Glass bottle
Specific Gravity 4.75
Assay Percent Range 98% min. (Argentometry)
Density 4.7500g/mL
Linear Formula BiCl3
Fieser 07,24
Merck Index 15, 1264
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in acids, soluble in abs.alcohol, acetone and ethyl acetate
Formula Weight 315.33
Percent Purity ≥97%
Chemical Name or Material Bismuth(III) chloride, Anhydrous
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Bismuth(III) chloride, anhydrous
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.