missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) chloride, anhydrous, 99.9% (metals basis)

Product Code. 11303327
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303327 100 g 100g
11393317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303327 Supplier Thermo Scientific Alfa Aesar Supplier No. 017115.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol

Catalysts for organic reactions, preparation of Bi salts.Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-60-2
Molecular Weight (g/mol) 315.33
MDL Number MFCD00003461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
Synonym bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth lll chloride
PubChem CID 24591
IUPAC Name trichlorobismuthane
SMILES Cl[Bi](Cl)Cl

Specifications

Boiling Point ca 430°C (sublimation)
Melting Point 230°C to 232°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (Metals basis)
Molecular Formula BiCl3
Solubility Information Soluble in HCl,HNO3,acetone,ethyl acetate. Insoluble in alcohol. Decomposed by water to BiOCl.Soluble in hydrochloric acid,nitric acid,diethyl ether,ethyl acetate and acetone. Insoluble in alcohol.
Formula Weight 315.34
Odor Irritating
Sensitivity Hygroscopic
Density 4.75
Chemical Name or Material Bismuth(III) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bismuth(III) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.