missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) Chloride, Extra Pure, SLR, Fisher Chemical

Product Code. 11483673 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483673 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483673 Supplier Fisher Chemical Supplier No. B/3110/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 447°C
Melting Point 230°C
CAS 7787-60-2
Color Gray
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula BiCl3
MDL Number 3461
InChI Key JHXKRIRFYBPWGE-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Bi+3]
IUPAC Name bismuth(3+) trichloride
Molecular Weight (g/mol) 315.33
Formula Weight 315.33g/mol
Packaging Glass Jar
Show More Show Less
compliance-icons
Product Identifier
  • Bismuth(III) chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.