missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) isopropoxide, Thermo Scientific Chemicals

Product Code. 11314988
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314988 5 g 5g
11304988 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314988 Supplier Thermo Scientific Alfa Aesar Supplier No. 043938.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15049-67-9 | Molecular Formula: C9H21BiO3 | Molecular Weight: 386.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15049-67-9
Molecular Formula C9H21BiO3
Molecular Weight (g/mol) 386.244
MDL Number MFCD00799065
InChI Key KNPRLIQQQKEOJN-UHFFFAOYSA-N
Synonym bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate
PubChem CID 22648700
IUPAC Name bismuth;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3]

Specifications

Color Gray
Linear Formula Bi[OCH(CH3)2]3
Quantity 5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 386.24
Physical Form Powder
Chemical Name or Material Bismuth (III) isopropoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.