missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, 98+%, pure

Product Code. 15209584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15209584 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15209584 Supplier Thermo Scientific Acros Supplier No. 192620050
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1890°C
Melting Point 825.0°C
CAS 1304-76-3
Color Gray to White
Physical Form Powder
Quantity 5 kg
Specific Gravity 8.9
Assay Percent Range 98.0 to 102%
Density 8.9000g/mL
Molecular Formula Bi2O3
Linear Formula Bi2O3
MDL Number MFCD00003462
Fieser 01,59
Merck Index 15, 1273
Synonym Bismuth trioxide
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]
IUPAC Name dibismuth(3+) trioxidandiide
Molecular Weight (g/mol) 465.96
Formula Weight 465.95
Percent Purity ≥98%
Grade Pure
Chemical Name or Material Bismuth(III) oxide, 99.9999%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.