Learn More
Bismuth(III) oxide, 99.975% (metals basis)
Description
Used as Adhesives and sealant chemicals, flame retardants, pigments, propellants and blowing agents. It is also used as solvents. Other uses are in batteries, electrical and electronic products, explosive materials ink, toner, and colorant products, paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | ca 1890°C |
| Melting Point | 817°C |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.975% (Metals basis) |
| Solubility Information | Insoluble in water. Soluble in acids. |
| Formula Weight | 465.96 |
| Odor | Odorless |
| Density | 8.9 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.