missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) oxide, NanoArc™, 99.5+%

Product Code. 15442387
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442387 500 g 500g
15422387 25 g 25g
15432387 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442387 Supplier Thermo Scientific Alfa Aesar Supplier No. 045582.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol

Bismuth(III) oxide is used in solid-oxide fuel cells. It is also used in optical glasses, to replace lead oxide in whitewares, luxes, varistor formulations and ceramic capacitor formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • X-ray opaque particle
  • NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ∼1890°C
Melting Point 817°C
Color Yellow
Physical Form Powder
Quantity 500 g
Merck Index 14,1273
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 465.96
Percent Purity ≥99.5%
Odor Odorless
Density 8.9 g/mL
Chemical Name or Material Bismuth (III) oxide NanoArc™ BI-7300
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.