Learn More
Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)
Description
It is used as a precursor to the preparation of other compounds of bismuth, chemical reagents. It has specialized uses in optical glass, flame-retardant paper, in glaze formulations as a substitute for lead oxides. It is used as solid electrolyte because of its high thermal expansion property. It is used in radio frequency shielding, compound for catheters and other medical tubing products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | ∼1890°C |
| Melting Point | 817°C |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 250 g |
| Density | 8.9 g/mL |
| Merck Index | 14,1273 |
| Solubility Information | Soluble in acids. Insoluble in water & organic solvents. |
| Formula Weight | 465.96 |
| Percent Purity | 99.9995% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.