missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) Oxide, TRC

Product Code. 30344117
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344117 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344117 Supplier Toronto Research Chemicals Supplier No. TRCB4973435G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bismuth(III) oxide
CAS 1304-76-3
Quantity 5 g
Molecular Formula Bi2O3
Synonym Bismuth oxide (Bi2O3), B 18401, Bismuth oxide, Bismuth oxide (BiO1.5), Bismuth sesquioxide, Bismuth sesquioxide (Bi2O3), Bismuth trioxide, Bismuth yellow, Bismuth(3+) oxide, Bismuth(III) oxide, Bismuthous oxide, C.I. 77160, Dibismuth trioxide, NanoArc Bismuth Oxide, Nanotek, Tomatec 42-970A
InChI Formula InChI=1S/2Bi.3O
SMILES O=[Bi]O[Bi]=O
IUPAC Name oxo(oxobismuthanyloxy)bismuthane
Molecular Weight (g/mol) 465.959
Formula Weight 465.9455
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.