missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) Subnitrate, 98%, Honeywell Fluka™

Product Code. 15613210 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
5g
Product Code. Quantity Packaging unitSize
15613210 5 g Poly Bottle 5g
15693200 100 g Poly Bottle 100g
15603210 500 g Poly Bottle 500g
3 options
This item is not returnable. View return policy

Product Code. 15613210

Brand: Honeywell-Fluka 3106465G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.98

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-85-4
MDL Number MFCD00064844

Specifications

Melting Point >500°C
Quantity 5 g
Linear Formula Bi5O(OH)9(NO3)4
UN Number UN1477
Formula Weight 1461.99g/mol
Percent Purity 98%
Packaging Poly Bottle
Density 5.29g/cm3
Chemical Name or Material Bismuth(Iii) Subnitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.