missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) tert-pentyloxide, 97+%

Product Code. 11349567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349567 5 g 5g
11339567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349567 Supplier Thermo Scientific Alfa Aesar Supplier No. 041203.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 124687-44-1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124687-44-1
MDL Number MFCD00151666
Synonym Bismuth(III) tert-amylate; Bismuth(III) tert-pentoxide

Specifications

Melting Point 180°C (decomposition)
Color Gray
Boiling Point 63°C (at 10-4mmHg)(sublimation)
Linear Formula Bi(OC5H11)3
Quantity 5 g
UN Number UN3181
Formula Weight 470.41
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Bismuth(III) tert-pentyloxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.