missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) Trifluoromethanesulfonate (3:1), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Bismuth(III) Trifluoromethanesulfonate (3:1) |
| CAS | 88189-03-1 |
| Molecular Formula | C3BiF9O9S3 |
| Quantity | 5 g |
| Synonym | Bismuth Triflate, Bismuth Tris(trifluoromethanesulfonate), Bismuth(3+) Triflate, Bismuth(3+) Trifluoromethanesulfonate, Bismuth(III) Triflate, Bismuth(III) Trifluoromethanesulfonate, Trifluoromethanesulfonic Acid Bismuth(3+) Salt |
| InChI Formula | InChI=1S/3CHF3O3S.Bi/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3 |
| SMILES | [Bi+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
| IUPAC Name | bismuth;trifluoromethanesulfonate |
| Molecular Weight (g/mol) | 656.19 |
| Formula Weight | 655.8365 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto