missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Nitrate Pentahydrate, Certified AR for Analysis, Fisher Chemical

Product Code. 10714561 Shop All Fisher Chemical Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10714561 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10714561 Supplier Fisher Chemical Supplier No. B/3200/48
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 80°C
Melting Point 30°C
CAS 10035-06-0
Color White
Physical Form Solid
Quantity 100 g
Molecular Formula BiN3O9·5H2O
MDL Number 149157
Formula Weight 485.07g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.