missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Subnitrate, TRC

Product Code. 30341930
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30341930 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30341930

Brand: Toronto Research Chemicals TRCB49728510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bismuth Subnitrate
CAS 1304-85-4
Molecular Formula Bi H O2 . 4 Bi H2 N O5
Quantity 10 g
Synonym Bismuth subnitrate, Bismuth hydroxide nitrate oxide (Bi5O(OH)9(NO3)4) (8CI), Basic Bismuth Nitrate, Bismuth Magistery, Bismuth Oxynitrate, Bismuth Subnitrate, Bismuth Subnitricum, Bismuth white, Bismuthyl Nitrate, Blanc de Fard, C.I. 77169, C.I. Pigment White 17, Cosmetic White, Flake White, Magistery of bismuth, Novismuth, OrbeSeal, Pigment White 17, Roter, Snowcal 5SW, Spanish White, Vicalin, Vikaline
InChI Formula InChI=1S/5Bi.4NO3.9H2O.O/c;;;;;4*2-1(3)4;;;;;;;;;;/h;;;;;;;;;9*1H2;/q+1;4*+3;4*-1;;;;;;;;;;/p-9
SMILES O[Bi]=O.O[Bi](O)O[N+](=O)[O-].O[Bi](O)O[N+](=O)[O-].O[Bi](O)O[N+](=O)[O-].O[Bi](O)O[N+](=O)[O-]
IUPAC Name dihydroxybismuthanyl nitrate;hydroxy(oxo)bismuthane
Molecular Weight (g/mol) 1461.99
Formula Weight 1461.8728
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.