missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth Subnitrate, TRC
Specifications
Specifications
| Chemical Name or Material | Bismuth Subnitrate |
| CAS | 1304-85-4 |
| Molecular Formula | Bi H O2 . 4 Bi H2 N O5 |
| Quantity | 25 g |
| Synonym | Bismuth subnitrate, Bismuth hydroxide nitrate oxide (Bi5O(OH)9(NO3)4) (8CI), Basic Bismuth Nitrate, Bismuth Magistery, Bismuth Oxynitrate, Bismuth Subnitrate, Bismuth Subnitricum, Bismuth white, Bismuthyl Nitrate, Blanc de Fard, C.I. 77169, C.I. Pigment White 17, Cosmetic White, Flake White, Magistery of bismuth, Novismuth, OrbeSeal, Pigment White 17, Roter, Snowcal 5SW, Spanish White, Vicalin, Vikaline |
| InChI Formula | InChI=1S/5Bi.4NO3.9H2O.O/c;;;;;4*2-1(3)4;;;;;;;;;;/h;;;;;;;;;9*1H2;/q+1;4*+3;4*-1;;;;;;;;;;/p-9 |
| SMILES | O[Bi]=O.O[Bi](O)O[N+](=O)[O-].O[Bi](O)O[N+](=O)[O-].O[Bi](O)O[N+](=O)[O-].O[Bi](O)O[N+](=O)[O-] |
| IUPAC Name | dihydroxybismuthanyl nitrate;hydroxy(oxo)bismuthane |
| Molecular Weight (g/mol) | 1461.99 |
| Formula Weight | 1461.8728 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?