missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Bisoprolol-d7 Hemifumarate (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Bisoprolol-d7 Hemifumarate |
| CAS | 1310012-16-8 |
| Molecular Formula | 2 [C18 D7 H24 N O4] . C4 H4 O4 |
| Quantity | 0.01 g |
| Synonym | 1-(1,1,1,2,3,3,3-Heptadeuteriopropan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol hemifumarate, Bisoprolol-d7 hemifumarate, Bisoprolol-D7 (N-isopropyl-D7) hemifumarate, 1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-2-propanol (2 E)-2-butenedioate (2:1) |
| InChI Formula | InChI=1 S/2C18H31NO4.C4H4O4/c2*1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4;5-3(6)1-2-4(7)8/h2*5-8,14-15,17,19-20 H,9-13H2,1-4H3;1-2 H,(H,5,6)(H,7,8)/b;;2-1+/i2*1D3,2D3,14 D; |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NCC(O)COc1ccc(COCCOC(C)C)cc1)C([2 H])([2 H])[2 H].[2 H]C([2 H])([2 H])C([2 H])(NCC(O)COc2ccc(COCCOC(C)C)cc2)C([2 H])([2 H])[2 H].OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;1-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol |
| Molecular Weight (g/mol) | 781.0445 |
| Formula Weight | 780.5495 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction