missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A Diglycidyl Ether, TRC

Product Code. 30344499
Change view
Click to view available options
Quantity:
50 mg
50 g
Unit Size:
50g
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344499 50 mg 50mg
30329855 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30344499 Supplier Toronto Research Chemicals Supplier No. TRCB51950050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Bis-(4-glycidyloxyphenyl)propane
CAS 1675-54-3
Purity Grade Notes HPLC
Molecular Formula C21 H24 O4
Quantity 50 mg
Synonym BADGE, Propane, 2,2-bis[p-(2,3-epoxypropoxy)phenyl]- (6CI,8CI), 2,2'-[(1-Methylethylidene)bis(4,1-phenyleneoxymethylene)]bis[oxirane], 2,2-Bis(4-glycidoxyphenyl)propane, 2,2-Bis(4-glycidyloxyphenyl)propane, 2,2-Bis(4-hydroxyphenyl)propane diglycidyl ether, 2,2-Bis(4'-glycidoxyphenyl)propane, 2,2-Bis(4'-glycidyloxyphenyl)propane, 2,2-Bis(p-glycidyloxyphenyl)propane, 2,2-Bis(p-hydroxyphenyl)propane diglycidyl ether, 2,2-Bis[4-(2,3-epoxypropoxy)phenyl]propane, 2,2-Bis[4-(2,3-epoxypropyloxy)phenyl]propane, 2,2-Bis[p-(2,3-epoxypropoxy)phenyl]propane, 2,2-Di(4-glycidyloxyphenyl)propane, 4,4'-Bis(2,3-epoxypropoxy)diphenyldimethylmethane, 4,4'-Isopropylidenebis[1-(2,3-epoxypropoxy)benzene], 4,4'-Isopropylidenediphenol diglycidyl ether, Badge (crosslinking agent), Bis(4-hydroxyphenyl)dimethylmethane diglycidyl ether, Bisphenol A bisglycidyl ether, Bisphenol A bisoxiranyl ether, Bisphenol A diglycidyl ether, DGEBA, Dian diglycidyl ether, Diglycidyl bisphenol A ether, Diglycidyl diphenylolpropane ether, Diglycidylbisphenol A, Diomethane diglycidyl ether, NSC 5022
InChI Formula InChI=1S/C21H24O4/c1-21(2,15-3-7-17(8-4-15)22-11-19-13-24-19)16-5-9-18(10-6-16)23-12-20-14-25-20/h3-10,19-20H,11-14H2,1-2H3
SMILES CC(C)(c1ccc(OCC2CO2)cc1)c3ccc(OCC4CO4)cc3
IUPAC Name 2-[[4-[2-[4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane
Molecular Weight (g/mol) 340.41
Formula Weight 340.1675
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.