missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol F acetate propionate, epoxy curative, EC-298

Product Code. 15447295
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447295 100 g 100g
15437295 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447295 Supplier Thermo Scientific Alfa Aesar Supplier No. 046724.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Well suited for use in epoxy/(meth)acrylate hybrid thermoset compositions | CAS: 1071466-53-9 | C18H18O4 | 298.338 g/mol

Bisphenol F acetate propionate does not impede free radical cure, thermoset adhesives, used as curative for epoxy/(meth) acrylate hybrids, hybrid epoxy/free radical thermosets. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071466-53-9
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.338
MDL Number MFCD22380705
InChI Key JARUUDOWTQBKKU-UHFFFAOYSA-N
Synonym 4-4-acetyloxy phenyl methyl phenyl propanoate, bisphenol f acetate propionate, epoxy curative, ec-298
PubChem CID 59317436
IUPAC Name [4-[(4-acetyloxyphenyl)methyl]phenyl] propanoate
SMILES CCC(=O)OC1=CC=C(C=C1)CC2=CC=C(C=C2)OC(=O)C

Specifications

Color Yellow
Physical Form Pale yellow liquid
Density 1.13
Solubility Information Soluble in ethanol,ether,chloroform,alkali; slightly soluble in DMSO; insoluble in carbon disulfide.
Quantity 100 g
Formula Weight 298.34
Flash Point 150°C (302°F)
Chemical Name or Material Bisphenol F acetate propionate, Epoxy curative, EC-298
compliance-icons
Product Identifier
  • Bisphenol F acetate propionate, epoxy curative, EC-298
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Bisphenol F acetatepropionate, epoxy curative

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.