missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol F Bis(2,3-dihydroxypropyl) Ether, TRC

Product Code. 30329820
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329820 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329820 Supplier Toronto Research Chemicals Supplier No. TRCB519550250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisphenol F Bis(2,3-dihydroxypropyl) Ether
CAS 72406-26-9
Purity Grade Notes HPLC
Molecular Formula C19 H24 O6
Quantity 250 mg
Synonym BFDGE.2H2O, 3,3'-[Methylenebis(4,1-phenyleneoxy)]bis[1,2-propanediol], Bisphenol F bis(2,3-dihydroxypropyl) ether
InChI Formula InChI=1S/C19H24O6/c20-10-16(22)12-24-18-5-1-14(2-6-18)9-15-3-7-19(8-4-15)25-13-17(23)11-21/h1-8,16-17,20-23H,9-13H2
SMILES OCC(O)COc1ccc(Cc2ccc(OCC(O)CO)cc2)cc1
IUPAC Name 3-[4-[[4-(2,3-dihydroxypropoxy)phenyl]methyl]phenoxy]propane-1,2-diol
Molecular Weight (g/mol) 348.39
Formula Weight 348.1573
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.