missing translation for 'onlineSavingsMsg'
Learn More

Bispyrazolone, TRC

Product Code. 30005783
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005783 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30005783 Supplier Toronto Research Chemicals Supplier No. TRCB5242001G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7477-67-0
Molecular Formula C20 H18 N4 O2
Molecular Weight (g/mol) 346.38
Synonym 2,2',4,4'-Tetrahydro-5,5'-dimethyl-2,2'-diphenyl-[4,4'-Bi-3H-pyrazole]-3,3'-dione, 3,3'-Dimethyl-1,1'-diphenyl-[4,4'-bi-2-pyrazoline]-5,5'-dione, 4,4'-Bis(1-phenyl-3-methyl-5-pyrazolone), NSC 37393, NSC 401001, Edaravone dimer
IUPAC Name 5-methyl-4-(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)-2-phenyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1C2C(=NN(C2=O)c3ccccc3)C)c4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H18N4O2/c1-13-17(19(25)23(21-13)15-9-5-3-6-10-15)18-14(2)22-24(20(18)26)16-11-7-4-8-12-16/h3-12,17-18H,1-2H3
Formula Weight 346.143
Chemical Name or Material Bispyrazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.