missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Biuret, 97%, extra pure

Product Code. 11945461 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11945461

Brand: Thermo Scientific Chemicals 106521000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-19-0 | C2H5N3O2 | 103.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.08
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea
SMILES C(=O)(N)NC(=O)N

Specifications

Melting Point 187°C to 194°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex N)
Packaging Glass bottle
Linear Formula H2NCONHCONH2
Quantity 100 g
Beilstein 03,7
Merck Index 15,131
Solubility Information Solubility in water: 2g/100mL water (25°C). Other solubilities: freely soluble in alcohols,slightly soluble in diethyl ether
Formula Weight 103.08
Percent Purity 97%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Biuret
Show More Show Less
compliance-icons
Product Identifier
  • Biuret
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.