missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BIX-02188, 95%

Product Code. 17181126 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17181126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17181126 Supplier Thermo Scientific™ Supplier No. 467971000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material BIX-02188
CAS 334949-59-6
Synonym (3 Z)-3-[[[3-[(Dimethylamino)methyl]phenyl]amino]phenylmethylene]-2,3-dihydro-2-oxo-1 H-indole-6-carboxamide
Quantity 100 mg
Molecular Formula C25 H24 N4 O2
Formula Weight 412.49
Physical Form Solid
Product Type Reagent
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.