missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BIX NHE1 inhibitor

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BIX NHE1 inhibitor is a potent and selective NHE1 inhibitor (IC50 = 31 nM). Selective for NHE1 over NHE2 and NHE3. Prevents ischemic damage in an ischemia reperfusion injury isolated rat heart model ex vivo. Prevents phenylephrine -induced cardiomyocyte hypertrophy in vitro, and attenuates cardiac hypertrophy and left ventricular dysfunction postinfarction in rats. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-(1-Acetyl-4-piperidinyl)-N-(aminoiminomethyl)-3-(trifluoromethyl)benzamide hydrochloride |
| CAS | 1422252-46-7 |
| Quantity | 10 mg |
| Target | Na+/Ca2+ Exchanger Inhibitors |
| Molecular Formula | C16H19F3N4O2.HCl |
| Formula Weight | 392.8 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction